siRNA Details
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175114.3 | NP_780323.2 | 5 | 659 | ||
GenBank mRNA | AK029123.1 | - | 6 | 1057 | ||
AK041436.1 | - | 6 | 1027 | |||
AK052264.1 | - | 6 | 1046 | |||
AK122426.1 | - | 6 | 749 | |||
BC046285.1 | - | 5 | 649 | |||
BC058971.1 | - | 5 | 601 | 6832507 | ||
BC116262.1 | - | 5 | 626 | |||
BC116263.1 | - | 5 | 626 | |||
BC129961.1 | - | 5 | 626 | |||
BC129962.1 | - | 5 | 626 |
