siRNA Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025985.4 | NP_080261.2 | 4 | 632 | ||
GenBank mRNA | AK012539.1 | - | 4 | 539 | ||
AK013902.1 | - | 4 | 513 | |||
AK083029.1 | - | 4 | 575 | |||
AK167472.1 | - | 4 | 498 | |||
BC065083.1 | - | 5 | 726 | |||
BC096474.1 | - | 749 | 4503077 | |||
DV657669.1 | - | 4 | 632 |
