siRNA Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025985.4 | NP_080261.2 | 4 | 711 | ||
GenBank mRNA | AK012539.1 | - | 4 | 618 | ||
AK013902.1 | - | 4 | 592 | |||
AK167472.1 | - | 4 | 577 | |||
BC065083.1 | - | 5 | 805 | |||
BC096474.1 | - | 828 | 4503077 | |||
DV657669.1 | - | 4 | 711 |
