siRNA Details
Gene Symbol: 2610301B20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026005.3 | NP_080281.3 | 5 | 575 | ||
GenBank mRNA | AK011950.1 | - | 5 | 504 | ||
AK146358.1 | - | 5 | 496 | |||
BC058777.1 | - | 5 | 465 | 5397173 | ||
BM948390.1 | - | 5 | 447 | |||
CX220600.1 | - | 5 | 575 |
