siRNA Details
Gene Symbol: 3110009F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014028.1 | - | 2, 3 | 325 | ||
AK014095.1 | - | 2, 3 | 300 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110009F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014028.1 | - | 2, 3 | 325 | ||
AK014095.1 | - | 2, 3 | 300 |