siRNA Details
Gene Symbol: 5730559C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028872.3 | NP_083148.3 | 4 | 668 | ||
GenBank mRNA | AK017848.1 | - | 3 | 308 | ||
BC052416.1 | - | 4 | 652 | 6402346 | ||
BC053100.1 | - | 4 | 652 | 6402346 |
