siRNA Details
Gene Symbol: 1700054K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006783.1 | - | 2 | 86 | ||
AK018861.1 | - | 2 | 86 | |||
AK018961.1 | - | 2 | 86 | |||
AK018979.1 | - | 2 | 86 | |||
BY707289.1 | - | 2 | 86 |
