siRNA Details
Gene Symbol: 1110008F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026124.3 | NP_080400.1 | 1 | 254 | ||
GenBank mRNA | AK003568.1 | - | 1 | 249 | ||
AK031002.1 | - | 1 | 232 | |||
AK078178.1 | - | 1 | 248 | |||
AK155559.1 | - | 1 | 193 | |||
BC019445.1 | - | 1 | 192 | 5125733 |
