siRNA Details
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 12 | 1490 | ||
NM_023341.3 | NP_075830.2 | 12 | 1588 | |||
GenBank mRNA | AJ278735.1 | - | 12 | 1468 | ||
AK004177.1 | - | 5 | 473 | |||
AK004791.1 | - | 12 | 1530 | |||
AK014605.2 | - | 12 | 1530 | |||
AK191635.1 | - | 3 | 213 | |||
AK192575.1 | - | 3 | 214 | |||
AK194890.1 | - | 3 | 213 | |||
AK196005.1 | - | 3 | 214 | |||
AK202430.1 | - | 3 | 213 | |||
AK207786.1 | - | 3 | 213 | |||
AK217974.1 | - | 4 | 328 | |||
BC030937.1 | - | 12 | 1536 | 4922892 | ||
U31629.1 | - | 10 | 1129 |
