siRNA Details
Gene Symbol: Fam120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024203.3 | NP_077165.2 | 2 | 1260 | ||
GenBank mRNA | AK016568.1 | - | 2 | 1228 | ||
AK028911.1 | - | 2 | 1190 | |||
AK050319.1 | - | 1 | 975 | |||
AK076185.1 | - | 2 | 1226 | |||
AK087438.1 | - | 2 | 1207 | |||
AK132211.1 | - | 2 | 1238 | |||
AK220425.1 | - | 2 | 1222 | |||
BC003200.1 | - | 1 | 166 | |||
BC066088.1 | - | 2 | 1305 | |||
BC085163.1 | - | 2 | 1258 | 30618952 | ||
DQ873694.1 | - | 2 | 1241 |
