siRNA Details
Gene Symbol: Mgat4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162368.1 | NP_001155840.1 | 6 | 1446 | ||
NM_001162369.1 | NP_001155841.1 | 6 | 1319 | |||
NM_001205098.1 | NP_001192027.1 | 8 | 1859 | |||
NM_026243.4 | NP_080519.2 | 4 | 1341 | |||
GenBank mRNA | AK018574.1 | - | 4 | 1340 | ||
AK018672.1 | - | 4 | 1344 | |||
AK032009.1 | - | 6 | 1322 | |||
AK033482.1 | - | 4 | 1342 | |||
AK033643.1 | - | 1 | 596 | |||
BC046987.1 | - | 4 | 1329 | 6475851 |
