siRNA Details
Gene Symbol: Mgat4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162368.1 | NP_001155840.1 | 5 | 499 | ||
NM_001162369.1 | NP_001155841.1 | 5 | 372 | |||
NM_001205098.1 | NP_001192027.1 | 7 | 912 | |||
NM_026243.4 | NP_080519.2 | 3 | 394 | |||
GenBank mRNA | AK018574.1 | - | 3 | 393 | ||
AK018672.1 | - | 3 | 397 | |||
AK032009.1 | - | 5 | 373 | |||
AK033482.1 | - | 3 | 395 | |||
BB611341.1 | - | 5 | 500 | |||
BC046987.1 | - | 3 | 382 | 6475851 | ||
CD774691.1 | - | 7 | 680 |
