siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 2 | 268 | ||
GenBank mRNA | AK008162.1 | - | 2 | 218 | ||
AK014790.1 | - | 2 | 213 | |||
AK086546.1 | - | 2 | 161 | |||
AK089102.1 | - | 2 | 232 | |||
BC053004.1 | - | 2 | 205 | |||
BC137692.1 | - | 2 | 268 | 8860711 |
