siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 1 | 141 | ||
GenBank mRNA | AK008162.1 | - | 1 | 91 | ||
AK012910.1 | - | 105 | ||||
AK014790.1 | - | 1 | 86 | |||
AK086546.1 | - | 1 | 34 | |||
AK089102.1 | - | 1 | 105 | |||
BC053004.1 | - | 1 | 78 | |||
BC137692.1 | - | 1 | 141 | 8860711 |
