siRNA Details
Gene Symbol: 4930548H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026296.3 | NP_080572.1 | 2 | 887 | ||
GenBank mRNA | AK016070.1 | - | 2 | 879 | ||
AK016690.1 | - | 2 | 888 | |||
BC057946.1 | - | 1 | 1983 |
Gene Symbol: 4930566F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016234.1 | - | 2 | 443 |
