siRNA Details
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042499.1 | NP_001035964.1 | 3 | 193 | ||
GenBank mRNA | AK011280.1 | - | 3 | 188 | ||
AK016099.1 | - | 3 | 202 | |||
AK028195.1 | - | 3 | 187 | |||
AK028269.1 | - | 3 | 179 | |||
AK156982.1 | - | 3 | 184 | |||
BC050194.1 | - | 3 | 174 | 30022531 |
