siRNA Details
Gene Symbol: Pcnxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026327.3 | NP_080603.2 | 9 | 2402 | ||
GenBank mRNA | AK029996.1 | - | 9 | 2359 | ||
AK136802.1 | - | 9 | 2377 | |||
AK160852.1 | - | 8 | 1622 | |||
BC019573.1 | - | 1 | 105 |
