siRNA Details
Gene Symbol: Reg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026328.2 | NP_080604.2 | 2, 3 | 226 | ||
GenBank mRNA | AK008049.1 | - | 2, 3 | 228 | ||
AK008438.1 | - | 2, 3 | 229 | |||
AK160329.1 | - | 2, 3 | 223 | |||
AK162316.1 | - | 2, 3 | 227 | |||
BC019465.1 | - | 2, 3 | 209 | 4216795 |
