siRNA Details
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026386.1 | NP_080662.1 | 2 | 171 | ||
GenBank mRNA | AK002692.1 | - | 2 | 171 | ||
AK005470.1 | - | 2 | 183 | |||
AK010572.1 | - | 2 | 168 | |||
AK075929.1 | - | 2 | 168 | |||
AK145840.1 | - | 2 | 222 | |||
AK153525.1 | - | 2 | 168 | |||
BC006960.1 | - | 2 | 171 | 2812557 | ||
CT010229.1 | - | 2 | 156 |
