siRNA Details
Gene Symbol: Snap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144521.2 | NP_653104.1 | 3 | 946 | ||
GenBank mRNA | AK028053.1 | - | 3 | 962 | ||
AK049330.1 | - | 3 | 990 | |||
AK075640.1 | - | 3 | 985 | |||
AK081883.1 | - | 3 | 962 | |||
AK189462.1 | - | 1 | 38 | |||
AK211035.1 | - | 1 | 57 | |||
BC023182.1 | - | 3 | 946 | 4949809 |
