siRNA Details
Gene Symbol: Murc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026509.3 | NP_080785.2 | 1 | 449 | ||
GenBank mRNA | AK009073.1 | - | 1 | 444 | ||
AK009691.1 | - | 1 | 406 | |||
BC089601.1 | - | 1 | 449 | 6788090 | ||
EU487254.1 | - | 1 | 341 |
