siRNA Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 2 | 392 | ||
GenBank mRNA | AB232609.1 | - | 1 | 23 | ||
AJ968955.1 | - | 2 | 247 | |||
AJ968956.1 | - | 2 | 247 | |||
AJ968957.1 | - | 2 | 247 | |||
AJ968958.1 | - | 2 | 247 | |||
AJ968959.1 | - | 2 | 247 | |||
AJ968960.1 | - | 2 | 247 | |||
AK002704.1 | - | 2 | 264 | |||
AK013410.1 | - | 2 | 245 | |||
AK133963.1 | - | 2 | 292 | |||
AK135751.1 | - | 2 | 281 | |||
AK160859.1 | - | 2 | 392 | |||
AK165812.1 | - | 2 | 239 | |||
AK168551.1 | - | 2 | 249 | |||
AK168867.1 | - | 2 | 261 | |||
AK169396.1 | - | 2 | 263 | |||
BC009085.1 | - | 2 | 75 | 2649852 | ||
BC025139.1 | - | 2 | 263 | 3980228 | ||
BC056648.1 | - | 2 | 265 | 5034519 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053589, XM_006234050, XM_006234049, BC158585, M83680.
Gene Ontology Categories:
