siRNA Details
Gene Symbol: Vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081041.1 | NP_001074510.1 | 6, 7 | 1672 | ||
GenBank mRNA | AK137155.1 | - | 6, 7 | 1682 | ||
AK146211.1 | - | 6, 7 | 1681 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081041.1 | NP_001074510.1 | 6, 7 | 1672 | ||
GenBank mRNA | AK137155.1 | - | 6, 7 | 1682 | ||
AK146211.1 | - | 6, 7 | 1681 |