siRNA Details
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026825.3 | NP_081101.3 | 31, 32 | 3011 | ||
GenBank mRNA | AK043591.1 | - | 17, 18 | 1609 | ||
AY437876.1 | - | 31, 32 | 3016 | |||
BC012229.1 | - | 10, 11 | 1040 | |||
BC150819.1 | - | 30, 31 | 2925 |
