siRNA Details
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026963.5 | NP_081239.1 | 4, 5 | 1004 | ||
GenBank mRNA | AK007657.1 | - | 4, 5 | 493 | ||
AK146824.1 | - | 4, 5 | 1071 | |||
AK210899.1 | - | 4, 5 | 334 | |||
BC023174.1 | - | 4, 5 | 1004 | 4952948 | ||
DQ058008.1 | - | 3, 4 | 329 |
