siRNA Details
Gene Symbol: Odf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027019.3 | NP_081295.2 | 7 | 743 | ||
GenBank mRNA | AK005883.1 | - | 7 | 854 | ||
BC104105.1 | - | 7 | 743 | 40043157 | ||
BC104106.1 | - | 7 | 743 | 40043160 | ||
BC106806.1 | - | 7 | 743 | 40042632 |
