siRNA Details
Gene Symbol: 1700008O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027049.1 | NP_081325.1 | 5 | 541 | ||
GenBank mRNA | AK005784.1 | - | 5 | 542 | ||
BC127243.1 | - | 3 | 657 | |||
BC127244.1 | - | 3 | 657 | |||
BF467486.1 | - | 2 | 382 |
