siRNA Details
Gene Symbol: 1700030J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027103.2 | NP_081379.1 | 3 | 442 | ||
GenBank mRNA | AK006553.1 | - | 3 | 447 | ||
AK030075.1 | - | 3 | 443 | |||
AK031529.1 | - | 3 | 327 | |||
BC049630.1 | - | 3 | 371 | |||
BC100501.1 | - | 3 | 462 | 30917058 | ||
BC137750.1 | - | 3 | 380 | 8860769 | ||
BC137751.1 | - | 3 | 380 | 8860770 |
