siRNA Details
Gene Symbol: 2310061I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033630.1 | NP_001028802.1 | 4 | 863 | ||
GenBank mRNA | AK010000.1 | - | 1 | 159 | ||
AK038622.1 | - | 3 | 1715 | |||
AK145087.1 | - | 4 | 817 | |||
AK172070.1 | - | 4 | 780 | |||
BC058418.1 | - | 1 | 108 | |||
BC100355.1 | - | 3 | 492 | 30527188 | ||
BC117035.1 | - | 3 | 523 | |||
BC117039.1 | - | 3 | 523 | |||
BC144744.1 | - | 3 | 523 | |||
BQ043979.1 | - | 1 | 110 | |||
BQ444983.1 | - | 3 | 306 |
