siRNA Details
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252447.1 | NP_001239376.1 | 8 | 979 | ||
NM_027260.3 | NP_081536.2 | 8 | 979 | |||
GenBank mRNA | AF513620.1 | - | 7 | 643 | ||
AK012664.1 | - | 8 | 671 | |||
AK154485.1 | - | 8 | 692 | |||
AK186520.1 | - | 2 | 137 | |||
BC013520.1 | - | 7 | 653 | 4236402 |
