siRNA Details
Gene Symbol: Lrch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081255.1 | NP_001074724.1 | 18 | 1982 | ||
GenBank mRNA | AK134026.1 | - | 8 | 614 | ||
AK155550.1 | - | 18 | 1980 | |||
BC151058.1 | - | 18 | 1960 | 9087971 | ||
BC157924.1 | - | 17 | 1852 | 9088097 | ||
BC172144.1 | - | 16 | 1702 | 9054814 |
