siRNA Details
Gene Symbol: Cd209g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027343.3 | NP_081619.3 | 8 | 710 | ||
GenBank mRNA | AK010071.1 | - | 7 | 671 | ||
AM286437.1 | - | 8 | 713 | |||
BC118542.1 | - | 1 | 3235 |
