siRNA Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027352.4 | NP_081628.3 | 3 | 378 | ||
GenBank mRNA | AK002515.1 | - | 4 | 457 | ||
AK033413.1 | - | 3 | 312 | |||
AK044691.1 | - | 263 | ||||
AK077153.1 | - | 3 | 254 | |||
AK077683.1 | - | 3 | 313 | |||
AK077706.1 | - | 3 | 245 | |||
AK148337.1 | - | 3 | 312 | |||
AK151843.1 | - | 3 | 260 | |||
AK152475.1 | - | 3 | 260 | |||
AK152544.1 | - | 3 | 260 | |||
AK152603.1 | - | 3 | 288 | |||
AK153049.1 | - | 3 | 260 | |||
AK153555.1 | - | 3 | 312 | |||
AK167823.1 | - | 3 | 312 | |||
AK168928.1 | - | 4 | 508 | |||
AK169382.1 | - | 3 | 260 | |||
AK210658.1 | - | 2 | 136 | |||
BC005600.1 | - | 3 | 251 | 3482776 | ||
BC016455.1 | - | 2 | 103 | 4234343 | ||
BC024982.1 | - | 3 | 264 | |||
BU707592.1 | - | 3 | 378 |
