siRNA Details
Gene Symbol: Bhlhb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098222.1 | NP_001091692.1 | 5 | 1953 | ||
NM_198161.2 | NP_937804.1 | 5 | 1963 | |||
GenBank mRNA | AK012577.1 | - | 1 | 447 | ||
AK214737.1 | - | 1 | 25 | |||
BC059871.1 | - | 1 | 1318 | 6809364 |
