siRNA Details
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027375.2 | NP_081651.2 | 6 | 1167 | ||
GenBank mRNA | AB093232.1 | - | 6 | 1169 | ||
AK045701.1 | - | 6 | 1174 | |||
BC027339.1 | - | 6 | 1149 | |||
BC139007.1 | - | 6 | 1163 | 8862029 | ||
BC145495.1 | - | 6 | 1236 |
