siRNA Details
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 8 | 1421 | ||
GenBank mRNA | AK005465.1 | - | 8 | 1373 | ||
BC002308.1 | - | 3 | 497 | |||
BC051550.1 | - | 6 | 977 | |||
BC095941.1 | - | 8 | 1353 | 4167912 | ||
BM936626.1 | - | 4 | 682 | |||
EF668007.1 | - | 8 | 1277 |
