siRNA Details
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 3 | 419 | ||
GenBank mRNA | AK005465.1 | - | 3 | 374 | ||
BB867118.1 | - | 3 | 433 | |||
BC095941.1 | - | 3 | 351 | 4167912 | ||
CK023294.1 | - | 3 | 361 | |||
EF668007.1 | - | 3 | 278 |
