siRNA Details
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183288.3 | NP_899111.2 | 3 | 1263 | ||
GenBank mRNA | AK017636.1 | - | 3 | 955 | ||
BC138292.1 | - | 1 | 719 | 8861311 | ||
BC138293.1 | - | 1 | 719 | 8861312 |
Gene Symbol: Arhgap27os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052773.1 | - | 2 | 503 |
