siRNA Details
Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027468.1 | NP_081744.1 | 9 | 1315 | ||
GenBank mRNA | AK017670.1 | - | 3 | 545 | ||
BC047389.1 | - | 8 | 1301 | |||
BC088735.1 | - | 9 | 1320 | |||
BC100404.1 | - | 9 | 1309 | 30471377 |
