siRNA Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164233.1 | NP_001157705.1 | 10, 11 | 1823 | ||
NM_027584.1 | NP_081860.1 | 10, 11 | 1823 | |||
GenBank mRNA | AK014872.1 | - | 10, 11 | 1824 | ||
AK133206.1 | - | 11, 12 | 1912 | |||
BC112423.1 | - | 10, 11 | 1793 | 40062055 | ||
BC113131.1 | - | 10, 11 | 1793 | 40062053 |
