siRNA Details
Gene Symbol: 4931406C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199484.1 | NP_001186413.1 | 9 | 1191 | ||
NM_001199485.1 | NP_001186414.1 | 9 | 1087 | |||
NM_133732.3 | NP_598493.1 | 8 | 1050 | |||
GenBank mRNA | AK016432.1 | - | 8 | 946 | ||
AK088123.1 | - | 9 | 1051 | |||
AK133192.1 | - | 9 | 1190 | |||
AK147073.1 | - | 9 | 1061 | |||
BC011540.1 | - | 8 | 953 | 2648220 | ||
BC016078.1 | - | 8 | 999 | 4489072 | ||
BC025213.1 | - | 8 | 951 | |||
CT010296.1 | - | 8 | 864 |
