siRNA Details
Gene Symbol: 5530601H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080296.1 | - | 6 | 650 | ||
AK084653.1 | - | 6 | 655 | |||
AK084980.1 | - | 6 | 634 | |||
AK085098.1 | - | 6 | 627 | |||
AK085485.1 | - | 6 | 621 | |||
AK164650.1 | - | 5 | 581 | |||
BC055463.1 | - | 6 | 529 |
