siRNA Details
Gene Symbol: Zfp949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142943.1 | NP_001136415.1 | 6 | 2166 | ||
NM_001163061.1 | NP_001156533.1 | 5 | 2020 | |||
GenBank mRNA | AK137745.1 | - | 1 | 16 | ||
BC141009.1 | - | 6 | 2003 | 9055621 | ||
BC141010.1 | - | 5 | 1857 | 9055622 |
