siRNA Details
Gene Symbol: Ubiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027873.2 | NP_082149.1 | 1 | 656 | ||
GenBank mRNA | AK004550.1 | - | 1 | 656 | ||
AK034787.1 | - | 1 | 683 | |||
AK078442.2 | - | 1 | 657 | |||
BC015303.1 | - | 1 | 364 | 4241204 | ||
BC071203.1 | - | 1 | 645 | 3667534 |
