siRNA Details
Gene Symbol: 1700007B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164235.1 | NP_001157707.1 | 10 | 1300 | ||
NM_027944.1 | NP_082220.1 | 9 | 1182 | |||
GenBank mRNA | AK005693.1 | - | 9 | 1183 | ||
AK077102.1 | - | 11 | 1038 | |||
BC117767.1 | - | 10 | 1203 | 40090978 | ||
BC117768.1 | - | 9 | 1085 | 40090984 |
