siRNA Details
Gene Symbol: Ppapdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028000.1 | NP_082276.1 | 6, 7 | 674 | ||
GenBank mRNA | AK007542.1 | - | 3, 4 | 1278 | ||
AK009455.1 | - | 5, 6 | 1284 | |||
AK144593.1 | - | 6, 7 | 659 | |||
BC023111.1 | - | 6, 7 | 621 | |||
BC039626.1 | - | 5, 6 | 489 | |||
BC062173.1 | - | 6, 7 | 607 | |||
BC089556.1 | - | 6, 7 | 576 | |||
DV661598.1 | - | 3, 4 | 324 |
