siRNA Details
Gene Symbol: 2310047M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028005.3 | NP_082281.2 | 1 | 425 | ||
GenBank mRNA | AK009886.1 | - | 1 | 468 | ||
AK151629.1 | - | 1 | 427 | |||
AK169875.1 | - | 1 | 406 |
Gene Symbol: 9330160F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137386.1 | - | 1 | 282 |
