siRNA Details
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028026.2 | NP_082302.2 | 2, 3 | 162 | ||
GenBank mRNA | AK004708.1 | - | 1, 2 | 600 | ||
AK010448.1 | - | 2, 3 | 128 | |||
AK034740.1 | - | 2, 3 | 126 | |||
AK049132.1 | - | 2, 3 | 160 | |||
AK169377.1 | - | 1, 2 | 599 | |||
AK169527.1 | - | 2, 3 | 162 | |||
AK220207.1 | - | 2, 3 | 131 | |||
BC028870.1 | - | 2, 3 | 114 | 4037614 | ||
BC068122.1 | - | 1, 2 | 589 | |||
BC087940.1 | - | 2, 3 | 121 | 30302907 |
