siRNA Details
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028026.2 | NP_082302.2 | 5, 6 | 393 | ||
GenBank mRNA | AK004708.1 | - | 4, 5 | 831 | ||
AK010448.1 | - | 5, 6 | 359 | |||
AK034740.1 | - | 5, 6 | 357 | |||
AK049132.1 | - | 5, 6 | 391 | |||
AK169377.1 | - | 4, 5 | 830 | |||
AK169527.1 | - | 5, 6 | 393 | |||
BC028870.1 | - | 5, 6 | 346 | 4037614 | ||
BC068122.1 | - | 4, 5 | 820 | |||
BC087940.1 | - | 5, 6 | 352 | 30302907 |
