siRNA Details
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028026.2 | NP_082302.2 | 6 | 463 | ||
GenBank mRNA | AK004708.1 | - | 5 | 901 | ||
AK010448.1 | - | 6 | 429 | |||
AK034740.1 | - | 6 | 427 | |||
AK049132.1 | - | 6 | 461 | |||
AK169377.1 | - | 5 | 900 | |||
AK169527.1 | - | 6 | 463 | |||
AK220207.1 | - | 7 | 498 | |||
BC028870.1 | - | 6 | 416 | 4037614 | ||
BC068122.1 | - | 5 | 890 | |||
BC087940.1 | - | 6 | 422 | 30302907 |
